ENA448969102 Enamine

N-methyl-2-[(4-methylpiperidin-4-yl)formamido]acetamide hydrochloride

  • MDL number MFCD21605967
  • Cas Number : 1361118-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213263
Availability details for
Quantity
  • Availability
  • Add to Cart