ENA448969105 Enamine

4,6-dichloro-2-(difluoromethyl)quinoline

  • MDL number MFCD17240871
  • Cas Number : 1343235-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213744
Availability details for
Quantity
  • Availability
  • Add to Cart