ENA448969107 Enamine

[(3-bromothiophen-2-yl)methyl][2-(morpholin-4-yl)ethyl]amine

  • MDL number MFCD16093892
  • Cas Number : 1247757-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213159
Availability details for
Quantity
  • Availability
  • Add to Cart