ENA448969109 Enamine

2-ethylpyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD28505988
  • Cas Number : 1803595-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213758
Availability details for
Quantity
  • Availability
  • Add to Cart