ENA448969111 Enamine

3-[ethyl(methyl)carbamoyl]benzene-1-sulfonyl chloride

  • MDL number MFCD11623045
  • Cas Number : 1094841-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213277
Availability details for
Quantity
  • Availability
  • Add to Cart