ENA448969112 Enamine

5-(aminomethyl)-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD21302495
  • Cas Number : 1495375-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213799
Availability details for
Quantity
  • Availability
  • Add to Cart