ENA448969113 Enamine

2-(ethylamino)-5-nitrobenzene-1-sulfonamide

  • MDL number MFCD16662360
  • Cas Number : 127781-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213800
Availability details for
Quantity
  • Availability
  • Add to Cart