ENA448969114 Enamine

3-hydroxy-5,5-dimethylhexanoic acid

  • MDL number MFCD17254946
  • Cas Number : 149650-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213801
Availability details for
Quantity
  • Availability
  • Add to Cart