ENA448969122 Enamine

1-[(3-fluorophenyl)methyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD17525738
  • Cas Number : 1490700-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213809
Availability details for
Quantity
  • Availability
  • Add to Cart