ENA448969123 Enamine

4-chloro-N-methyl-3-(trifluoromethyl)aniline

  • MDL number MFCD11132239
  • Cas Number : 42265-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213812
Availability details for
Quantity
  • Availability
  • Add to Cart