ENA448969124 Enamine

[1-(2-methoxyphenyl)propyl](methyl)amine

  • MDL number MFCD10032966
  • Cas Number : 104178-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213815
Availability details for
Quantity
  • Availability
  • Add to Cart