ENA448969128 Enamine

2-[(3-chlorophenyl)methyl]oxirane

  • MDL number MFCD22414043
  • Cas Number : 25642-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213819
Availability details for
Quantity
  • Availability
  • Add to Cart