ENA448969129 Enamine

{2-amino-1-[4-(2-methylpropyl)phenyl]ethyl}dimethylamine

  • MDL number MFCD23861883
  • Cas Number : 1465573-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213821
Availability details for
Quantity
  • Availability
  • Add to Cart