ENA448969131 Enamine

methyl (3R)-3-amino-3-(naphthalen-1-yl)propanoate hydrochloride

  • MDL number MFCD24410413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213823
Availability details for
Quantity
  • Availability
  • Add to Cart