ENA448969136 Enamine

5-(methoxymethyl)-1,2-oxazole-3-carbonitrile

  • MDL number MFCD21706427
  • Cas Number : 1463940-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213842
Availability details for
Quantity
  • Availability
  • Add to Cart