ENA448969142 Enamine

ethyl 2-chloro-3,3-dimethylbutanoate

  • MDL number MFCD16694322
  • Cas Number : 98551-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213859
Availability details for
Quantity
  • Availability
  • Add to Cart