ENA448969153 Enamine

3-[(2-aminophenyl)sulfanyl]-1-methylpyrrolidin-2-one

  • MDL number MFCD20285092
  • Cas Number : 1344249-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213921
Availability details for
Quantity
  • Availability
  • Add to Cart