ENA448969155 Enamine

(1S)-7-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD26585965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-213925
Availability details for
Quantity
  • Availability
  • Add to Cart