ENA448969157 Enamine

8-bromoimidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD20258809
  • Cas Number : 1289104-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213927
Availability details for
Quantity
  • Availability
  • Add to Cart