ENA448969158 Enamine

N-[2-(piperidin-3-yl)ethyl]acetamide hydrochloride

  • MDL number MFCD21606192
  • Cas Number : 1361113-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213928
Availability details for
Quantity
  • Availability
  • Add to Cart