ENA448969161 Enamine

2-[2-(sulfanylmethyl)phenyl]acetic acid

  • MDL number MFCD20657374
  • Cas Number : 913394-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213933
Availability details for
Quantity
  • Availability
  • Add to Cart