ENA448969163 Enamine

5-bromo-4-chloropyridine-2-carbonitrile

  • MDL number MFCD18257821
  • Cas Number : 1256790-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213947
Availability details for
Quantity
  • Availability
  • Add to Cart