ENA448969172 Enamine

benzyl 2-methanesulfonylacetate

  • MDL number MFCD25958774
  • Cas Number : 175465-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213962
Availability details for
Quantity
  • Availability
  • Add to Cart