ENA448969174 Enamine

methyl[3-(methylamino)propyl][(pyridin-4-yl)methyl]amine

  • MDL number MFCD14655521
  • Cas Number : 1249748-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213966
Availability details for
Quantity
  • Availability
  • Add to Cart