ENA448969177 Enamine

1,3-dimethyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD06120837
  • Cas Number : 88398-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-105
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213970
Availability details for
Quantity
  • Availability
  • Add to Cart