ENA448969178 Enamine

3-(3-aminopropyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD22070876
  • Cas Number : 1367990-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-213971
Availability details for
Quantity
  • Availability
  • Add to Cart