ENA448969210 Enamine

[1-(3-methoxyphenyl)propyl](methyl)amine

  • MDL number MFCD09761260
  • Cas Number : 941256-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214093
Availability details for
Quantity
  • Availability
  • Add to Cart