ENA448969212 Enamine

1-chloro-7-fluoro-6-methoxyisoquinoline

  • MDL number MFCD21876101
  • Cas Number : 630422-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214098
Availability details for
Quantity
  • Availability
  • Add to Cart