ENA448969213 Enamine

3-(4-bromo-1H-pyrazol-1-yl)propan-1-amine

  • MDL number MFCD03422574
  • Cas Number : 1000802-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214099
Availability details for
Quantity
  • Availability
  • Add to Cart