ENA448969215 Enamine

tert-butyl 4-(4-cyano-2-fluorophenoxy)piperidine-1-carboxylate

  • MDL number MFCD28506010
  • Cas Number : 696588-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214102
Availability details for
Quantity
  • Availability
  • Add to Cart