ENA448969229 Enamine

4-chloro-2,3-dihydro-1H-isoindole hydrochloride

  • MDL number MFCD11110370
  • Cas Number : 924304-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-214128
Availability details for
Quantity
  • Availability
  • Add to Cart