ENA448969246 Enamine

N,N-diethyl-2-imino-2,3-dihydro-1,3-benzothiazol-6-amine

  • MDL number MFCD31566996
  • Cas Number : 121436-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214163
Availability details for
Quantity
  • Availability
  • Add to Cart