ENA448969253 Enamine

3-[(2-amino-4-chlorophenyl)sulfanyl]-1-methylpyrrolidin-2-one

  • MDL number MFCD20285096
  • Cas Number : 1339603-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214184
Availability details for
Quantity
  • Availability
  • Add to Cart