ENA448969256 Enamine

1-(1-methyl-1H-1,2,3-triazol-5-yl)methanamine hydrochloride

  • MDL number MFCD28506020
  • Cas Number : 1781060-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214188
Availability details for
Quantity
  • Availability
  • Add to Cart