ENA448969258 Enamine

4-amino-1-{2-[(2-methoxyethyl)(methyl)amino]ethyl}pyrrolidin-2-one dihydrochloride

  • MDL number MFCD28505942
  • Cas Number : 1803612-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213335
Availability details for
Quantity
  • Availability
  • Add to Cart