ENA448969260 Enamine

2-(2-chlorophenyl)-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine hydrobromide

  • MDL number MFCD28506023
  • Cas Number : 1004527-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214194
Availability details for
Quantity
  • Availability
  • Add to Cart