ENA448969266 Enamine

tert-butyl 3-(cyanomethyl)-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD20230559
  • Cas Number : 1330766-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214226
Availability details for
Quantity
  • Availability
  • Add to Cart