ENA448969272 Enamine

N,2,2-triphenylethen-1-imine

  • MDL number MFCD00685317
  • Cas Number : 14181-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214307
Availability details for
Quantity
  • Availability
  • Add to Cart