ENA448969278 Enamine

1-{3-[(oxolan-3-yl)methoxy]pyridin-4-yl}methanamine

  • MDL number MFCD24158700
  • Cas Number : 1509515-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214315
Availability details for
Quantity
  • Availability
  • Add to Cart