ENA448969280 Enamine

3-{[2-(4-chlorophenyl)ethyl]amino}-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD19054332
  • Cas Number : 1410612-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214320
Availability details for
Quantity
  • Availability
  • Add to Cart