ENA448969281 Enamine

sodium 4-ethoxybenzene-1-sulfinate

  • MDL number MFCD24113276
  • Cas Number : 111243-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214321
Availability details for
Quantity
  • Availability
  • Add to Cart