ENA448969284 Enamine

1-(4,5,6,7-tetrahydro-1,2-benzoxazol-3-yl)methanamine

  • MDL number MFCD08701263
  • Cas Number : 893639-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214334
Availability details for
Quantity
  • Availability
  • Add to Cart