ENA448969288 Enamine

{N''-[2-(4-chlorophenyl)ethyl]carbamimidamido}methanimidamide hydrochloride

  • Cas Number : 110181-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214352
Availability details for
Quantity
  • Availability
  • Add to Cart