ENA448969289 Enamine

1-(2-methoxyphenyl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD17053891
  • Cas Number : 1483768-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214364
Availability details for
Quantity
  • Availability
  • Add to Cart