ENA448969293 Enamine

1-[3-(3,3-dimethylbutoxy)pyridin-4-yl]methanamine

  • MDL number MFCD24158710
  • Cas Number : 1515070-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214369
Availability details for
Quantity
  • Availability
  • Add to Cart