ENA448969294 Enamine

1-(4-amino-octahydro-1H-isoindol-2-yl)ethan-1-one

  • MDL number MFCD26881420
  • Cas Number : 1565012-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214370
Availability details for
Quantity
  • Availability
  • Add to Cart