ENA448969296 Enamine

1-methyl-1H-indene-2-carboxylic acid

  • MDL number MFCD00021226
  • Cas Number : 66130-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214372
Availability details for
Quantity
  • Availability
  • Add to Cart