ENA448969297 Enamine

5-nitro-2-(propylamino)benzene-1-sulfonamide

  • MDL number MFCD17433162
  • Cas Number : 1337888-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213344
Availability details for
Quantity
  • Availability
  • Add to Cart