ENA448969301 Enamine

(chloromethoxy)cyclobutane

  • MDL number MFCD19233804
  • Cas Number : 104620-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214398
Availability details for
Quantity
  • Availability
  • Add to Cart