ENA448969306 Enamine

2-({[4-fluoro-2-(trifluoromethyl)phenyl]methyl}amino)ethan-1-ol

  • MDL number MFCD24141320
  • Cas Number : 1516051-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214411
Availability details for
Quantity
  • Availability
  • Add to Cart